• USA Home
  • 442836 - 3-Methyl-L-histidine hydrate

EMAIL THIS PAGE TO A FRIEND
442836 Aldrich

3-Methyl-L-histidine hydrate

99%

  • CAS Number 207556-04-5

  • Empirical Formula (Hill Notation) C7H11N3O2 · xH2O

  • Molecular Weight 169.18 (anhydrous basis)

  •  EC Number 206-704-6

  •  PubChem Substance ID 329756490

Purchase

Properties

InChI Key   JVZHCTIDJLKMST-RGMNGODLSA-N
assay   99%
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?