• USA Home
  • 443352 - N-tert-Butyl-N-ethylisobutylamine

EMAIL THIS PAGE TO A FRIEND
443352 Sigma-Aldrich

N-tert-Butyl-N-ethylisobutylamine

98%

  • CAS Number 244191-68-2

  • Linear Formula (CH3)2CHCH2N(C2H5)C(CH3)3

  • Molecular Weight 157.30

  •  MDL number MFCD00274228

Purchase

Properties

assay   98%
refractive index   n20/D 1.424 (lit.)
bp   157 °C (lit.)
density   0.772 g/mL at 25 °C (lit.)
SMILES string   CCN(CC(C)C)C(C)(C)C
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
96.8 °F
Flash Point(C) 
36 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?