• USA Home
  • 444138 - Neodymium(III) perchlorate solution

EMAIL THIS PAGE TO A FRIEND
444138 Sigma-Aldrich

Neodymium(III) perchlorate solution

40 wt. % in H2O, 99.9%

Purchase

Properties

vapor pressure   0.12 psi ( 20 °C)
  1.68 psi ( 55 °C)
assay   99.9%
concentration   40 wt. % in H2O
density   1.436 g/mL at 25 °C
SMILES string   [Nd+3].[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O
Safety & Documentation

Safety Information

RIDADR 
UN 3098 5.1/PG 2

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?