• USA Home
  • 444383 - 1-(4-Nitrophenyl)-1H-imidazole

EMAIL THIS PAGE TO A FRIEND
444383 Sigma-Aldrich

1-(4-Nitrophenyl)-1H-imidazole

95%

  • CAS Number 2301-25-9

  • Empirical Formula (Hill Notation) C9H7N3O2

  • Molecular Weight 189.17

  •  MDL number MFCD00060491

  •  PubChem Substance ID 24868120

Purchase

Properties

assay   95%
mp   198-203 °C (lit.)
SMILES string   [O-][N+](=O)c1ccc(cc1)-n2ccnc2
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
NI7962000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?