• USA Home
  • 444901 - erythro-2-Anilino-1,2-diphenylethanol

EMAIL THIS PAGE TO A FRIEND
444901 Sigma-Aldrich

erythro-2-Anilino-1,2-diphenylethanol

95%

Purchase

Properties

assay   95%
mp   122-127 °C (lit.)
SMILES string   [H][C@@](O)(c1ccccc1)[C@]([H])(Nc2ccccc2)c3ccccc3
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?