• USA Home
  • 444928 - erythro-1,2-Diphenyl-2-(propylamino)ethanol

EMAIL THIS PAGE TO A FRIEND
444928 Sigma-Aldrich

erythro-1,2-Diphenyl-2-(propylamino)ethanol

98%

Purchase

Properties

assay   98%
optical activity   [α]23/D +1.4°, c = 1 in chloroform
mp   120-124 °C (lit.)
SMILES string   CCCN[C@@H]([C@@H](O)c1ccccc1)c2ccccc2
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?