• USA Home
  • 444979 - Trisodium triphenylphosphine-3,3′,3″-trisulfonate

EMAIL THIS PAGE TO A FRIEND
444979 Sigma-Aldrich

Trisodium triphenylphosphine-3,3′,3″-trisulfonate

96% (excludes the solvent of recrystallization)

Synonym: 3,3′,3″-Phosphinidynetris(benzenesulfonic acid) trisodium salt, Triphenylphosphine-3,3′,3′′-trisulfonic acid trisodium salt, Tris(3-sulfophenyl)phosphine trisodium salt

  • CAS Number 63995-70-0

  • Linear Formula P(C6H4SO3Na)3

  • Molecular Weight 568.42

  •  Beilstein/REAXYS Number 3584807

  •  EC Number 264-596-6

  •  MDL number MFCD00145472

Purchase

Properties

InChI Key   MYAJTCUQMQREFZ-UHFFFAOYSA-K
assay   96% (excludes the solvent of recrystallization)

Description

Application

Ligand for Rh- and Ru-based catalyst.

Safety & Documentation

Safety Information

WGK Germany 
1

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?