• USA Home
  • 445150 - Diethyl dithiophosphate potassium salt

EMAIL THIS PAGE TO A FRIEND
445150 Sigma-Aldrich

Diethyl dithiophosphate potassium salt

97%

Purchase

Properties

assay   97%
mp   187 °C (lit.)
SMILES string   [K+].CCOP([S-])(=S)OCC
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?