• USA Home
  • 446173 - 3-(Dimethoxymethylsilyl)-1-propanethiol

EMAIL THIS PAGE TO A FRIEND
446173 Sigma-Aldrich

3-(Dimethoxymethylsilyl)-1-propanethiol

95%

Synonym: (3-Mercaptopropyl)methyldimethoxysilane, Dimethoxy-(3-mercaptopropyl)methylsilane

  • CAS Number 31001-77-1

  • Linear Formula CH3Si(OCH3)2CH2CH2CH2SH

  • Molecular Weight 180.34

  •  Beilstein/REAXYS Number 2424867

  •  EC Number 250-426-8

  •  MDL number MFCD00053923

  •  PubChem Substance ID 24868228

Purchase

Properties

assay   95%
form   liquid
refractive index   n20/D 1.45 (lit.)
bp   96 °C/30 mmHg (lit.)
density   1 g/mL at 25 °C (lit.)
SMILES string   CO[Si](C)(CCCS)OC
InChI   1S/C6H16O2SSi/c1-7-10(3,8-2)6-4-5-9/h9H,4-6H2,1-3H3
InChI key   IKYAJDOSWUATPI-UHFFFAOYSA-N
Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
180 °F
Flash Point(C) 
82 °C
Protocols & Articles

Articles

Mesoporous Oxides and Their Applications to Hydrogen Storage

Solid-state hydrogen storage is attractive from a technological point of view, but has encountered tremendous challenges in terms of practical storage capacity and kinetics.1-3 Hydrogen sorption, whe...
Arlon J. Hunt1, Karl Gross2, Samuel S. Mao1
Material Matters Volume 4 Article 2
Keywords: Addition reactions, Adsorption, Applications, Catalysis, Chemical vapor deposition, Condensations, Deposition, Diffusion, Insecticides, Magnetic resonance spectroscopy, Material Matters, Methods, Microscopy, Nanomaterials, Photographs, Scanning electron microscopy, Size-exclusion chromatography, Sol-gel, Substitutions, Transmission electron microscopy

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?