• USA Home
  • 446629 - S-Hexyl-L-glutathione

EMAIL THIS PAGE TO A FRIEND
446629 Aldrich

S-Hexyl-L-glutathione

98%

Synonym: S-Hexyl-L-glutathione reduced

  • CAS Number 24425-56-7

  • Linear Formula HO2CCH(NH2)CH2CH2CONHCH[CH2SCH2(CH2)4CH3]CONHCH2CO2H

  • Molecular Weight 391.48

  •  Beilstein/REAXYS Number 5629635

  •  MDL number MFCD00056735

Purchase

Properties

assay   98%
mp   200-202 °C (lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

H6886 S-Hexylglutathione

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?