• USA Home
  • 447897 - Tri(ethylene glycol) dinonanoate

EMAIL THIS PAGE TO A FRIEND
447897 Sigma-Aldrich

Tri(ethylene glycol) dinonanoate

Purchase

Properties

refractive index   n20/D 1.446 (lit.)
bp   210 °C(lit.)
density   0.965 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?