• USA Home
  • 448338 - 16-Bromohexadecanoic acid

EMAIL THIS PAGE TO A FRIEND
448338 Sigma-Aldrich

16-Bromohexadecanoic acid

technical grade, 85%

Purchase

Properties

Related Categories Building Blocks, C13 to C42+, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
grade   technical grade
InChI Key   PFNCOYVEMJYEED-UHFFFAOYSA-N
assay   85%
impurities   <15% 16-[(16-bromo-1-oxohexadecyl)oxy]hexadecanoic acid
bp   214-217 °C/1 mmHg(lit.)
mp   68-71 °C (lit.)
  70-73 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?