• USA Home
  • 450448 - 4,4′-Ethylidenebisphenol

EMAIL THIS PAGE TO A FRIEND
450448 Sigma-Aldrich

4,4′-Ethylidenebisphenol

99%

Synonym: 1,1-Bis(4-hydroxyphenyl)ethane, Bisphenol E

Purchase

Properties

Related Categories Bisphenol and Sulfonyldiphenol Monomers, Materials Science, Monomers, Polymer Science
assay   99%
mp   123-127 °C (lit.)
SMILES string   CC(c1ccc(O)cc1)c2ccc(O)cc2
InChI   1S/C14H14O2/c1-10(11-2-6-13(15)7-3-11)12-4-8-14(16)9-5-12/h2-10,15-16H,1H3
InChI key   HCNHNBLSNVSJTJ-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?