• USA Home
  • 450618 - Diphenyl N-succinimidyl phosphate

EMAIL THIS PAGE TO A FRIEND
450618 Sigma-Aldrich

Diphenyl N-succinimidyl phosphate

97%

Synonym: SDPP

  • CAS Number 75513-55-2

  • Empirical Formula (Hill Notation) C16H14NO6P

  • Molecular Weight 347.26

  •  MDL number MFCD00274258

Purchase

Properties

assay   97%
mp   83-86 °C (lit.)
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?