• USA Home
  • 450839 - Dysprosium(III) chloride

EMAIL THIS PAGE TO A FRIEND
450839 Sigma-Aldrich

Dysprosium(III) chloride

anhydrous, powder, 99.9%

Synonym: Dysprosium chloride, Dysprosium trichloride

Purchase

Properties

grade   anhydrous
assay   99.9%
form   powder
mp   680 °C (lit.)
density   3.67 g/mL at 25 °C (lit.)
SMILES string   Cl[Dy](Cl)Cl
InChI   1S/3ClH.Dy/h3*1H;/q;;;+3/p-3
InChI key   BOXVSFHSLKQLNZ-UHFFFAOYSA-K
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
JW0700000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

325546 Dysprosium(III) chloride, anhydrous, powder, 99.99% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?