• USA Home
  • 450928 - Holmium(III) chloride hexahydrate

EMAIL THIS PAGE TO A FRIEND
450928 Sigma-Aldrich

Holmium(III) chloride hexahydrate

99.99%

Synonym: Holmium trichloride hexahydrate

Purchase

Properties

assay   99.99%
mp   165 °C (dec.) (lit.)
SMILES string   [H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].Cl[Ho](Cl)Cl
InChI   1S/3ClH.Ho.6H2O/h3*1H;;6*1H2/q;;;+3;;;;;;/p-3
InChI key   BUQFCXABQYNXLP-UHFFFAOYSA-K
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?