• USA Home
  • 451150 - Lithium tetrachlorogallate

EMAIL THIS PAGE TO A FRIEND
451150 Sigma-Aldrich

Lithium tetrachlorogallate

anhydrous, powder, ≥99.99% trace metals basis

Purchase

Properties

grade   anhydrous
assay   ≥99.99% trace metals basis
form   powder
mp   175 °C (lit.)
SMILES string   [Li+].Cl[Ga-](Cl)(Cl)Cl
InChI   1S/4ClH.Ga.Li/h4*1H;;/q;;;;+3;+1/p-4
InChI key   KMBVSAMILLNTNQ-UHFFFAOYSA-J

Description

Application

Shows promise as the electrolyte in rechargeable batteries.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 3260 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?