• USA Home
  • 452025 - cis-1,2,3-Triphenylaziridine

EMAIL THIS PAGE TO A FRIEND
452025 Sigma-Aldrich

cis-1,2,3-Triphenylaziridine

98%

  • CAS Number 7042-42-4

  • Empirical Formula (Hill Notation) C20H17N

  • Molecular Weight 271.36

  •  MDL number MFCD00674021

  •  PubChem Substance ID 24868673

Purchase

Properties

assay   98%
mp   98-102 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?