• USA Home
  • 452068 - trans-1-Isopropyl-2,3-diphenylaziridine

EMAIL THIS PAGE TO A FRIEND
452068 Sigma-Aldrich

trans-1-Isopropyl-2,3-diphenylaziridine

97%

Purchase

Properties

assay   97%
contains   1-3% sodium hydroxide as inhibitor
mp   64-67 °C (lit.)
SMILES string   CC(C)N1[C@H]([C@@H]1c2ccccc2)c3ccccc3
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?