• USA Home
  • 452734 - N-(1H-Benzotriazol-1-ylphenylmethyl)-3-pyridinecarboxamide

EMAIL THIS PAGE TO A FRIEND
452734 Sigma-Aldrich

N-(1H-Benzotriazol-1-ylphenylmethyl)-3-pyridinecarboxamide

97%

  • CAS Number 138768-28-2

  • Empirical Formula (Hill Notation) C19H15N5O

  • Molecular Weight 329.36

  •  MDL number MFCD00205103

  •  PubChem Substance ID 24868721

  •  NACRES NA.22

Purchase

Properties

Related Categories C-C Bond Formation, Chemical Synthesis, Other Synthetic Reagents for C-C Bond Formation, Synthetic Reagents
Quality Level   100
assay   97%
reaction suitability   reaction type: C-C Bond Formation
mp   184-188 °C (lit.)
SMILES string   O=C(NC(c1ccccc1)n2nnc3ccccc23)c4cccnc4
InChI   1S/C19H15N5O/c25-19(15-9-6-12-20-13-15)21-18(14-7-2-1-3-8-14)24-17-11-5-4-10-16(17)22-23-24/h1-13,18H,(H,21,25)
InChI key   BABUDAQOHSIBJR-UHFFFAOYSA-N

Description

Application

N-(1H-Benzotriazol-1-ylphenylmethyl)-3-pyridinecarboxamide can be used as an amidoalkylating reagent for the preparation of amidoalkylated products of aromatic and heterocyclic compounds.

Reactant for:
• Preparation of 3-substituted pyridinium salts

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Target organs 
Respiratory system
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Indoles, Purines, and Their Isosteres

Substituted indoles and purines have frequently been referred to as “privileged structures” since they are capable of binding to multiple receptors with high affinity, and thus have applications acro...
Keywords: Building blocks, Medicinal chemistry

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?