• USA Home
  • 452955 - 1-(4-Methoxyphenyl)-1H-pyrrole

EMAIL THIS PAGE TO A FRIEND
452955 Sigma-Aldrich

1-(4-Methoxyphenyl)-1H-pyrrole

97%

  • CAS Number 5145-71-1

  • Empirical Formula (Hill Notation) C11H11NO

  • Molecular Weight 173.21

  •  MDL number MFCD00128350

  •  PubChem Substance ID 24868742

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Pyrroles
InChI Key   WYFMHHMFUMBCGI-UHFFFAOYSA-N
assay   97%
mp   110-114 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis


Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?