• USA Home
  • 453641 - Fumaric acid monoethyl ester zinc salt

EMAIL THIS PAGE TO A FRIEND
453641 Sigma-Aldrich

Fumaric acid monoethyl ester zinc salt

97%

Purchase

Properties

assay   97%
mp   300 °C (dec.) (lit.)
SMILES string   [Zn++].[H]\C(=C(\[H])C(=O)OCC)C([O-])=O.[H]\C(=C(\[H])C(=O)OCC)C([O-])=O
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?