• USA Home
  • 453668 - Fumaric acid monoethyl ester magnesium salt

EMAIL THIS PAGE TO A FRIEND
453668 Sigma-Aldrich

Fumaric acid monoethyl ester magnesium salt

95%

Purchase

Properties

Related Categories Building Blocks, Carbonyl Compounds, Carboxylic Acid Salts, Chemical Synthesis, Organic Building Blocks More...
Quality Level   100
assay   95%
mp   169 °C (dec.) (lit.)
SMILES string   CCOC(=O)\C=C\C(=O)O[Mg]OC(=O)\C=C\C(=O)OCC
InChI   1S/2C6H8O4.Mg/c2*1-2-10-6(9)4-3-5(7)8;/h2*3-4H,2H2,1H3,(H,7,8);/q;;+2/p-2/b2*4-3+;
InChI key   ZHLRSUSHKGQBNV-SYWGCQIGSA-L

Description

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?