• USA Home
  • 453714 - (−)-Menthoxyacetyl chloride

EMAIL THIS PAGE TO A FRIEND
453714 Sigma-Aldrich

(−)-Menthoxyacetyl chloride

97%

  • CAS Number 15356-62-4

  • Empirical Formula (Hill Notation) C12H21ClO2

  • Molecular Weight 232.75

  •  MDL number MFCD00044947

  •  PubChem Substance ID 24868846

  •  NACRES NA.22

Purchase

Properties

Related Categories Acid Halides, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
Quality Level   100
assay   97%
optical activity   [α]25/D −10°, neat
refractive index   n20/D 1.469 (lit.)
density   1.033 g/mL at 25 °C (lit.)
SMILES string   CC(C)[C@@H]1CC[C@@H](C)C[C@H]1OCC(Cl)=O
InChI   1S/C12H21ClO2/c1-8(2)10-5-4-9(3)6-11(10)15-7-12(13)14/h8-11H,4-7H2,1-3H3/t9-,10+,11-/m1/s1
InChI key   VNMCKLVHDJADEB-OUAUKWLOSA-N

Description

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3265 8 / PGIII
WGK Germany 
WGK 3
Flash Point(F) 
235.4 °F - closed cup
Flash Point(C) 
113 °C - closed cup

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?