• USA Home
  • 454583 - Poly(tetrafluoroethylene-co-vinylidene fluoride-co-propylene)

EMAIL THIS PAGE TO A FRIEND
454583 Sigma-Aldrich

Poly(tetrafluoroethylene-co-vinylidene fluoride-co-propylene)

Purchase

Properties

composition   tetrafluoroethylene, 56%
  vinylidene fluoride, 27%
hardness   81 (Shore A)
viscosity   45(121 °C, Mooney, ML 1+10)(lit.)
density   1.6 g/mL at 25 °C (lit.)
SMILES string   CC=C.FC(F)=C.F\C(F)=C(\F)F

Description

Application

Gaskets and sealants. Injection, transfer and compression molding.

Features and Benefits

Resistant to new types of engine oils, transmission fluids and gear lubricants.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?