• USA Home
  • 454702 - Diethyl (dibromofluoromethyl)phosphonate

EMAIL THIS PAGE TO A FRIEND
454702 Sigma-Aldrich

Diethyl (dibromofluoromethyl)phosphonate

98%

Purchase

Properties

assay   98%
bp   212 °C (lit.)
density   1.756 g/mL at 25 °C (lit.)
SMILES string   CCOP(=O)(OCC)C(F)(Br)Br

Description

Application

Used to prepare monofluoro- and difluoro methylenephosphonates as phosphate mimics in bioorganic chemistry.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
217.4 °F
Flash Point(C) 
103 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?