• USA Home
  • 455237 - 4-(Diethylamino)benzophenone

EMAIL THIS PAGE TO A FRIEND
455237 Sigma-Aldrich

4-(Diethylamino)benzophenone

97%

Synonym: 4-(N ,N -Diethylamino)benzophenone, 4-(Diethylamino)benzophenone

  • CAS Number 18127-87-2

  • Linear Formula (C2H5)2NC6H4COC6H5

  • Molecular Weight 253.34

  •  MDL number MFCD00274308

  •  PubChem Substance ID 24869042

  •  NACRES NA.23

Purchase

Properties

Related Categories Amine Photochemical Coinitiators, Benzophenone, Materials Science, Organic Photoinitiators, Polymer Science,
assay   97%
mp   75-79 °C (lit.)
storage temp.   room temp
SMILES string   CCN(CC)c1ccc(cc1)C(=O)c2ccccc2
InChI   1S/C17H19NO/c1-3-18(4-2)16-12-10-15(11-13-16)17(19)14-8-6-5-7-9-14/h5-13H,3-4H2,1-2H3
InChI key   IDLJKTNBZKSHIY-UHFFFAOYSA-N

Description

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Target organs 
Respiratory system
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?