• USA Home
  • 455423 - Ammonium tetrakis(4-chlorophenyl)borate

EMAIL THIS PAGE TO A FRIEND
455423 Sigma-Aldrich

Ammonium tetrakis(4-chlorophenyl)borate

Purchase

Properties

mp   250 °C (dec.) (lit.)
SMILES string   [H][N+]([H])([H])[H].Clc1ccc(cc1)[B-](c2ccc(Cl)cc2)(c3ccc(Cl)cc3)c4ccc(Cl)cc4
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?