• USA Home
  • 455636 - 6,6′-Dihydroxy-5,5′-dimethoxy-[1,1′-biphenyl]-3,3′-dicarboxaldehyde

EMAIL THIS PAGE TO A FRIEND
455636 Sigma-Aldrich

6,6′-Dihydroxy-5,5′-dimethoxy-[1,1′-biphenyl]-3,3′-dicarboxaldehyde

technical grade, 90%

Synonym: Divanillin

  • CAS Number 2092-49-1

  • Linear Formula [HOC6H2(OCH3)(CHO)-]2

  • Molecular Weight 302.28

  •  MDL number MFCD00156888

Purchase

Properties

grade   technical grade
assay   90%
mp   315 °C (lit.)
absorption   λmax 357 nm

Description

Application

Antioxidant

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?