• USA Home
  • 455660 - Butyltriphenylphosphonium chloride

EMAIL THIS PAGE TO A FRIEND
455660 Sigma-Aldrich

Butyltriphenylphosphonium chloride

97%

Purchase

Properties

Related Categories C-C Bond Formation, Chemical Synthesis, Olefination, Synthetic Reagents, Wittig Reagents More...
assay   97%
mp   226-230 °C (lit.)
SMILES string   [Cl-].CCCC[P+](c1ccccc1)(c2ccccc2)c3ccccc3
InChI   1S/C22H24P.ClH/c1-2-3-19-23(20-13-7-4-8-14-20,21-15-9-5-10-16-21)22-17-11-6-12-18-22;/h4-18H,2-3,19H2,1H3;1H/q+1;/p-1
InChI key   MFIUDWFSVDFDDY-UHFFFAOYSA-M
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?