• USA Home
  • 456225 - trans-4-(2-Thienyl)-3-buten-2-one

EMAIL THIS PAGE TO A FRIEND
456225 Sigma-Aldrich

trans-4-(2-Thienyl)-3-buten-2-one

  • CAS Number 33603-63-3

  • Empirical Formula (Hill Notation) C8H8OS

  • Molecular Weight 152.21

  •  MDL number MFCD00053051

  •  PubChem Substance ID 24869139

  •  NACRES NA.22

Purchase

Properties

Related Categories Building Blocks, C7 to C9, Chemical Synthesis, Heterocyclic Building Blocks, Thiophenes More...
Quality Level   100
assay   ≥97.5%
mp   25-26 °C (lit.)
density   1.151 g/mL at 25 °C (lit.)
SMILES string   CC(=O)\C=C\c1cccs1
InChI   1S/C8H8OS/c1-7(9)4-5-8-3-2-6-10-8/h2-6H,1H3/b5-4+
InChI key   CIMALVIHZVKKPE-SNAWJCMRSA-N

Description

Packaging

5 mL in glass bottle

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?