EMAIL THIS PAGE TO A FRIEND
456837 Sigma-Aldrich

Bismuth(III) iodide

anhydrous, beads, −10 mesh, 99.999%

Synonym: Bismuth triiodide, Triiodobismuthane

Purchase

Properties

grade   anhydrous
InChI Key   KOECRLKKXSXCPB-UHFFFAOYSA-K
assay   99.999%
form   beads
particle size   −10 mesh
mp   408 °C (lit.)
density   5.78 g/mL at 25 °C(lit.)

Description

Application

Is used to make single crystals of alkyl bismuth diiodides which are useful intermediates in synthetic organic chemistry.

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3260 8 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?