• USA Home
  • 456888 - Poly(m-xylylenediamine-alt-epichlorohydrin), diamine terminated

EMAIL THIS PAGE TO A FRIEND
456888 Sigma-Aldrich

Poly(m-xylylenediamine-alt-epichlorohydrin), diamine terminated

average Mw ~670, average Mn ~325

Purchase

Properties

mol wt   average Mn ~325
  average Mw ~670
refractive index   n20/D 1.59 (lit.)
viscosity   8,000 cP(25 °C)(lit.)
solubility   lower alcohols and chloroform: soluble
  toluene, hexane and ethyl acetate: insoluble
density   1.14 g/mL at 25 °C (lit.)

Description

Application

Epoxy curing agent, chelating agent, lubricant and anti-corrosive additive.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?