EMAIL THIS PAGE TO A FRIEND
457930 Sigma-Aldrich

Scandium

chips, 99.9%

Synonym: Scandium element, Scandium-45

Purchase

Properties

InChI Key   SIXSYDAISGFNSX-UHFFFAOYSA-N
assay   99.9%
form   chips
resistivity   50.5 μΩ-cm, 0°C
bp   2836 °C (lit.)
mp   1540 °C (lit.)
density   2.99 g/mL at 25 °C (lit.)

Description

Application

Employed in solid state synthesis of unusual clusters, Sc19Br28Z4, (Z=Mn, Fe, Ru or Os). These clusters are of interest for their structure and magnetic properties.

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?