EMAIL THIS PAGE TO A FRIEND
459283 Sigma-Aldrich

Pentacontane

97%

  • CAS Number 6596-40-3

  • Linear Formula CH3(CH2)48CH3

  • Molecular Weight 703.34

  •  Beilstein/REAXYS Number 1807879

  •  EC Number 229-534-4

  •  MDL number MFCD00042907

Purchase

Properties

InChI Key   PFLUOWJPZLHUEA-UHFFFAOYSA-N
assay   97%
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?