EMAIL THIS PAGE TO A FRIEND
459763 Sigma-Aldrich

Tri-p-tolylamine

97%

Synonym: N ,N ,N -Tri(4-methylphenyl)amine, Tri(4-methylphenyl)amine, Tri-4-tolylamine, Tritolylamine

Purchase

Properties

Related Categories Hole Transport (HT) & Hole Injection Layer (HIL) Materials, Hole Transport Materials, Materials Science, OLED and PLED Materials, Organic and Printed Electronics More...
InChI Key   YXYUIABODWXVIK-UHFFFAOYSA-N
assay   97%
mp   114-118 °C (lit.)
storage temp.   2-8°C

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?