EMAIL THIS PAGE TO A FRIEND
461008 Aldrich

Potassium selenite

97%

Purchase

Properties

assay   97%
mp   875 °C(lit.)
Safety & Documentation

Safety Information

RIDADR 
UN 2630 6.1/PG 1
WGK Germany 
3
RTECS 
VS7200000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?