• USA Home
  • 461407 - (R)-(+)-2-Phenyl-1-propanol

EMAIL THIS PAGE TO A FRIEND
461407 Sigma-Aldrich

(R)-(+)-2-Phenyl-1-propanol

98%

Synonym: (R)-β-Methylphenethyl alcohol, (R)-2-Phenylpropanol

  • CAS Number 19141-40-3

  • Linear Formula CH3CH(C6H5)CH2OH

  • Molecular Weight 136.19

  •  Beilstein/REAXYS Number 2637521

  •  MDL number MFCD00145206

  •  PubChem Substance ID 24869852

  •  NACRES NA.22

Purchase

Properties

Related Categories Alcohols, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
Quality Level   200
assay   98%
optical activity   [α]22/D +17°, neat
refractive index   n20/D 1.526 (lit.)
bp   220 °C (lit.)
density   1 g/mL at 25 °C (lit.)
SMILES string   C[C@@H](CO)c1ccccc1
InChI   1S/C9H12O/c1-8(7-10)9-5-3-2-4-6-9/h2-6,8,10H,7H2,1H3/t8-/m0/s1
InChI key   RNDNSYIPLPAXAZ-QMMMGPOBSA-N

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
226.4 °F - closed cup
Flash Point(C) 
108 °C - closed cup
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?