• USA Home
  • 46197 - Farnesyl methyl ether

EMAIL THIS PAGE TO A FRIEND
46197 Sigma-Aldrich

Farnesyl methyl ether

technical, mixture of stereo isomers

  • CAS Number 15130-76-4

  • Empirical Formula (Hill Notation) C16H28O

  • Molecular Weight 236.39

  •  Beilstein/REAXYS Number 2042596

  •  EC Number 239-193-3

  •  MDL number MFCD00042862

  •  PubChem Substance ID 57650734

Purchase

Properties

Related Categories Building Blocks, C16 to C21, Chemical Synthesis, Ethers, Organic Building Blocks,
InChI Key   HZDZRGKULMGCJE-NCZFFCEISA-N
grade   technical
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?