• USA Home
  • 462160 - Disodium tris(1,2-benzenediolato-O,O′)silicate

EMAIL THIS PAGE TO A FRIEND
462160 Sigma-Aldrich

Disodium tris(1,2-benzenediolato-O,O′)silicate

90%

Purchase

Properties

assay   90%
impurities   1.6% methyl alcohol
mp   >400 °C (lit.)
SMILES string   [Na+].[Na+].O1c2ccccc2O[Si--]1345Oc6ccccc6O3.O4c7ccccc7O5
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3263 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?