• USA Home
  • 462543 - Potassium tetramethylcyclopentadienide

EMAIL THIS PAGE TO A FRIEND
462543 Sigma-Aldrich

Potassium tetramethylcyclopentadienide

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Other Ligands More...
InChI Key   IHPOCLHVUSDUSE-UHFFFAOYSA-N
mp   >240 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3393 4.2(4.3)
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?