• USA Home
  • 463329 - 5,5′-Methylenebis(2-hydroxy-4-methoxybenzophenone)

EMAIL THIS PAGE TO A FRIEND
463329 Sigma-Aldrich

5,5′-Methylenebis(2-hydroxy-4-methoxybenzophenone)

90%

Synonym: Bis(5-benzoyl-4-hydroxy-2-methoxyphenyl)methane

Purchase

Properties

Related Categories Materials Science, Polymer Additives, Polymer Science, Stabilizers
InChI Key   DCBNMBIOGUANTC-UHFFFAOYSA-N
assay   90%
bp   223-230 °C(lit.)
density   0.55 g/mL at 25 °C(lit.)

Description

Application

UV stabilizer

Packaging

10, 50 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?