EMAIL THIS PAGE TO A FRIEND
463426 Sigma-Aldrich

Trimethylboron-d9

electronic grade, 99.4% (CP), 99 atom % D

Synonym: Trimethylborane- d9

Purchase

Properties

Related Categories Boron, CVD and ALD Precursors by Metal, Chemical Synthesis, Materials Science, Micro/NanoElectronics,
grade   electronic grade
vapor density   2.3 (vs air)
InChI Key   WXRGABKACDFXMG-GQALSZNTSA-N
isotopic purity   99 atom % D
assay   99.4% (CP)
bp   −20.2 °C (lit.)
mp   −161.5 °C (lit.)
mass shift   M+9

Description

Packaging

5 g in stainless steel cylinder

Application

The deposition of trimethylboron-d9 derived boron-carbon films can result in increased device performance.

Recommended products

Stainless steel regulators Z527416 or Z527424 are recommended.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3160PIHB 2.1(2.3)
WGK Germany 
1
Protocols & Articles

Articles

High Purity Deposition Gases

High Purity CVD Gases are used in the deposition of amorphous and epitaxial silicon, SiGe alloys, and dielectrics as well as the manufacture of integrated circuits and photovolatics. The gases are al...
Chemfiles Volume 4 Article 3
Keywords: Chemical vapor deposition, Deposition

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?