• USA Home
  • 463795 - Dysprosium(III) oxalate hydrate

EMAIL THIS PAGE TO A FRIEND
463795 Sigma-Aldrich

Dysprosium(III) oxalate hydrate

≥99.99% trace metals basis

  • CAS Number 58176-69-5

  • Linear Formula Dy2(C2O4)3 · xH2O

  • Molecular Weight 589.06 (anhydrous basis)

  •  MDL number MFCD00674080

  •  PubChem Substance ID 24870098

Purchase

Properties

assay   ≥99.99% trace metals basis
form   powder
reaction suitability   reagent type: catalyst
core: dysprosium
SMILES string   O.[Dy+3].[Dy+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
InChI   1S/3C2H2O4.2Dy.H2O/c3*3-1(4)2(5)6;;;/h3*(H,3,4)(H,5,6);;;1H2/q;;;2*+3;/p-6
InChI key   QSUALEGWIZCPCI-UHFFFAOYSA-H
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?