• USA Home
  • 464619 - (Indan-1,3-diylidene)dimalononitrile

EMAIL THIS PAGE TO A FRIEND
464619 Sigma-Aldrich

(Indan-1,3-diylidene)dimalononitrile

97%

Purchase

Properties

assay   97%
mp   250 °C (dec.) (lit.)
SMILES string   N#C\C(C#N)=C1\CC(=C(\C#N)C#N)\c2ccccc12
InChI   1S/C15H6N4/c16-6-10(7-17)14-5-15(11(8-18)9-19)13-4-2-1-3-12(13)14/h1-4H,5H2
InChI key   HBZYYOYCJQHAEL-UHFFFAOYSA-N

Description

Application

Intermediate for the strong electron-withdrawing acceptor moiety in the synthesis of a novel push-pull chromophore.

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?