• USA Home
  • 464694 - Silver(I) perrhenate

EMAIL THIS PAGE TO A FRIEND
464694 Sigma-Aldrich

Silver(I) perrhenate

for synthesis

Purchase

Properties

Related Categories Essential Chemicals, Materials Science, Metal and Ceramic Science, Research Essentials, Salts,
grade   for synthesis
form   powder
mp   430 °C (lit.)
density   7.05 g/mL at 25 °C (lit.)
SMILES string   [Ag+].[O-][Re](=O)(=O)=O
InChI   1S/Ag.4O.Re/q+1;;;;-1;
InChI key   JRISYOCHUNKJAV-UHFFFAOYSA-N

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3260 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Single- & multi-channel pipettes from BRAND
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?