• USA Home
  • 465763 - 1,3-Diphenyl-2-thiobarbituric acid

EMAIL THIS PAGE TO A FRIEND
465763 Sigma-Aldrich

1,3-Diphenyl-2-thiobarbituric acid

97%

  • CAS Number 35221-12-6

  • Empirical Formula (Hill Notation) C16H12N2O2S

  • Molecular Weight 296.34

  •  MDL number MFCD00552520

  •  PubChem Substance ID 24870306

Purchase

Properties

assay   97%
drug control   regulated under CDSA - not available from Sigma-Aldrich Canada
mp   248 °C (dec.) (lit.)
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?