EMAIL THIS PAGE TO A FRIEND
466263 Aldrich

Cadmium

pellets, 6.35-6.35 mm, 99.999%

  • CAS Number 7440-43-9

  • Empirical Formula (Hill Notation) Cd

  • Molecular Weight 112.41

  •  Beilstein/REAXYS Number 8137359

  •  EC Number 231-152-8

  •  MDL number MFCD00010914

Purchase

Properties

InChI Key   BDOSMKKIYDKNTQ-UHFFFAOYSA-N
assay   99.999%
form   pellets
resistivity   7.27 μΩ-cm, 22°C
weight   1.57 g (pellet)
particle size   6.35-6.35 mm
bp   765 °C (lit.)
mp   320.9 °C (lit.)
density   8.65 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

202886 Cadmium, shot, 3 mm, 99.999% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?