• USA Home
  • 467316 - Dysprosium(III) carbonate hydrate

EMAIL THIS PAGE TO A FRIEND
467316 Aldrich

Dysprosium(III) carbonate hydrate

99.99% trace metals basis

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Dysprosium, Dysprosium Salts, Essential Chemicals,
InChI Key   MTSRAACGOZNKPN-UHFFFAOYSA-H
assay   99.99% trace metals basis
form   powder and chunks

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?