• USA Home
  • 467324 - Praseodymium(III) carbonate hydrate

EMAIL THIS PAGE TO A FRIEND
467324 Sigma-Aldrich

Praseodymium(III) carbonate hydrate

99.99%

  • CAS Number 54397-87-4

  • Linear Formula Pr2(CO3)3 · xH2O

  • Molecular Weight 461.84 (anhydrous basis)

  •  MDL number MFCD00149954

  •  PubChem Substance ID 24870400

Purchase

Properties

assay   99.99%
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?